missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium Chloride, 99.99%, (Optical Grade I), MP Biomedicals

Solutions are clear and colorless suitable for density gradient work, nucleic acid and protein purification

Supplier:  MP Biomedicals Inc 0216004191

 View more versions of this product

Catalog No. ICN16004191



Description

Description

Specifications

Specifications

Cesium Chloride
1 kg
ClCs
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
[Cl-].[Cs+]
168.36
CHEBI:63039
Optical I
7647-17-8
99.99%
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
caesium(1+) chloride
24293
168.4

Chemical Identifiers

7647-17-8
168.36
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
caesium(1+) chloride
ClCs
MFCD00010955
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
CHEBI:63039
[Cl-].[Cs+]
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions