missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium(IV) ammonium nitrate, 98+%, Thermo Scientific Chemicals

Cerium(IV) ammonium nitrate, >-98%, H8CeN8O18, CAS Number-16774-21-3, Ammonium cerium(IV) nitrate; CAN, 1000g, Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S., 240-827-6, 548.23, H272-H290-H302-H314-H317-H335

$66.33 - $606.68

Chemical Identifiers

CAS 16774-21-3
Molecular Formula H8CeN8O18
MDL Number MFCD00151121
Synonym Ammonium cerium(IV) nitrate; CAN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1288218
View Documents Promotion Details
Thermo Scientific Chemicals
A1288218
50 g
Each for $66.33
 
AAA1288230
View Documents Promotion Details
Thermo Scientific Chemicals
A1288230
250 g
Each for $192.56
 
AAA128820B
View Documents Promotion Details
Thermo Scientific Chemicals
A128820B
1000 g
Each for $606.68
 
Description

Description

Cerium(IV) ammonium nitrate is widely utilized as an oxidant for many functional groups like alcohols, phenols and ethers as well as C-H bonds. It acts as a deprotection reagent for alcohol in organic synthesis. It is useful in the synthesis of heterocycles like quinoxaline derivatives which is used in dyes, organic semiconductors and DNA cleaving agents. It is an active component of chrome etchant, which is used in the production of photomasks and liquid crystal displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerium(IV) ammonium nitrate is widely utilized as an oxidant for many functional groups like alcohols, phenols and ethers as well as C-H bonds. It acts as a deprotection reagent for alcohol in organic synthesis. It is useful in the synthesis of heterocycles like quinoxaline derivatives which is used in dyes, organic semiconductors and DNA cleaving agents. It is an active component of chrome etchant, which is used in the production of photomasks and liquid crystal displays.

Solubility
Soluble in water and alcohol.

Notes
Store in cool place. Hygroscopic. Incompatible with strong reducing agents and powdered metals.
Specifications

Chemical Identifiers

16774-21-3
H8CeN8O18
MFCD00151121
Ammonium cerium(IV) nitrate; CAN

Specifications

16774-21-3
H8CeN8O18
MFCD00151121
14,1992
Soluble in water and alcohol.
≥98%
Cerium(IV) ammonium nitrate
50 g
(NH4)2Ce(NO3)6
UN1477
Ammonium cerium(IV) nitrate; CAN
548.23
Pungent
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P390-P501c

H272-H290-H302-H314-H317-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

EINECSNumber : 240-827-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only