Greiner Bio-One CELLSTAR Suspension Culture Flask with Filter Cap
Ideal for suspension cultures, hybridoma and embryonic stem cells. Greiner Bio-One CELLSTAR Suspension Culture Flask with Filter Cap is free of detectable DNase, RNase and human DNA.
Supplier: Greiner Bio-One 658195
Description
- Features hydrophobic surface
- Graduation on the side
- Canted neck style
- Flask formats allows optimal access with pipettes
- Filter screw cap
Specifications
White | |
250 mL | |
Canted | |
5 | |
Yes | |
Filter |
HDPE, PET Filter | |
Polystyrene | |
120 | |
None | |
Sterile |