Learn More
Cefuroxime sodium salt, 95%, Thermo Scientific Chemicals
$204.53 - $661.27
Chemical Identifiers
CAS | 56238-63-2 |
---|---|
Molecular Formula | C16H15N4NaO8S |
Molecular Weight (g/mol) | 446.366 |
MDL Number | MFCD09878727 |
InChI Key | URDOHUPGIOGTKV-LNJPBTIASA-M |
PubChem CID | 133687825 |
IUPAC Name | sodium;(6S,7S)-3-(carbamoyloxymethyl)-7-[[(2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
SMILES | CON=C(C1=CC=CO1)C(=O)NC2C3N(C2=O)C(=C(CS3)COC(=O)N)C(=O)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6699903
|
Thermo Scientific Chemicals
J6699903 |
1 g |
Each for $204.53
|
|
|||||
AAJ6699906
|
Thermo Scientific Chemicals
J6699906 |
5 g |
Each for $661.27
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
56238-63-2 | |
446.366 | |
URDOHUPGIOGTKV-LNJPBTIASA-M | |
sodium;(6S,7S)-3-(carbamoyloxymethyl)-7-[[(2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
C16H15N4NaO8S | |
MFCD09878727 | |
133687825 | |
CON=C(C1=CC=CO1)C(=O)NC2C3N(C2=O)C(=C(CS3)COC(=O)N)C(=O)[O-].[Na+] |
Specifications
Cefuroxime Sodium Salt | |
56238-63-2 | |
C16H15N4NaO8S | |
14,1954 | |
URDOHUPGIOGTKV-LNJPBTIASA-M | |
sodium;(6S,7S)-3-(carbamoyloxymethyl)-7-[[(2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate | |
133687825 | |
95% | |
1 g |
Powder | |
240°C to 245°C | |
MFCD09878727 | |
Freely soluble in water | |
CON=C(C1=CC=CO1)C(=O)NC2C3N(C2=O)C(=C(CS3)COC(=O)N)C(=O)[O-].[Na+] | |
446.366 | |
446.37 | |
White to Yellow | |
Cefuroxime sodium salt |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
EINECSNumber : 260-073-1
RTECSNumber : XI0330000
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only