missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cefotiam dihydrochloride, Thermo Scientific Chemicals
A broad spectrum antibiotic
$460.19 - $686.84
Chemical Identifiers
CAS | 66309-69-1 |
---|---|
Molecular Formula | C18H27Cl2N9O5S3 |
Molecular Weight (g/mol) | 616.552 |
MDL Number | MFCD09878605 |
InChI Key | KHUCCKUOJMXYKH-BUWDDJPCSA-N |
PubChem CID | 70590037 |
IUPAC Name | (6R,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)acetyl]amino]-3-[[1-[2-(dimethylamino)ethyl]tetrazol-5-yl]sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride |
SMILES | CN(C)CCN1C(=NN=N1)SCC2=C(N3C(C(C3=O)NC(=O)CC4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ66529MF
|
Thermo Scientific Chemicals
J66529MF |
50 mg |
Each for $460.19
|
|
|||||
AAJ66529MC
|
Thermo Scientific Chemicals
J66529MC |
100 mg |
Each for $686.84
|
|
|||||
Description
A broad spectrum antibiotic
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
66309-69-1 | |
616.552 | |
KHUCCKUOJMXYKH-BUWDDJPCSA-N | |
(6R,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)acetyl]amino]-3-[[1-[2-(dimethylamino)ethyl]tetrazol-5-yl]sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride |
C18H27Cl2N9O5S3 | |
MFCD09878605 | |
70590037 | |
CN(C)CCN1C(=NN=N1)SCC2=C(N3C(C(C3=O)NC(=O)CC4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl |
Specifications
Cefotiam Dihydrochloride | |
66309-69-1 | |
MFCD09878605 | |
KHUCCKUOJMXYKH-BUWDDJPCSA-N | |
(6R,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)acetyl]amino]-3-[[1-[2-(dimethylamino)ethyl]tetrazol-5-yl]sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride | |
70590037 | |
White | |
>110°C (230°F) | |
Cefotiam dihydrochloride |
Powder | |
C18H27Cl2N9O5S3 | |
Soluble in water | |
CN(C)CCN1C(=NN=N1)SCC2=C(N3C(C(C3=O)NC(=O)CC4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl | |
616.552 | |
562.09 | |
50 mg | |
94°C |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 266-312-6
RTECSNumber : XI0367000
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only