missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cefdinir, 95%, Thermo Scientific Chemicals
$231.71 - $811.22
Chemical Identifiers
CAS | 91832-40-5 |
---|---|
Molecular Formula | C14H13N5O5S2 |
Molecular Weight (g/mol) | 395.408 |
MDL Number | MFCD00865030 |
InChI Key | RTXOFQZKPXMALH-GHXIOONMSA-N |
PubChem CID | 6915944 |
ChEBI | CHEBI:3485 |
IUPAC Name | (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
SMILES | C=CC1=C(N2C(C(C2=O)NC(=O)C(=NO)C3=CSC(=N3)N)SC1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6644703
|
Thermo Scientific Chemicals
J6644703 |
1 g |
N/A
|
|
|||||
AAJ6644706
|
Thermo Scientific Chemicals
J6644706 |
5 g |
N/A
|
|
|||||
Description
A broad spectrum antibiotic targeting both Gram-positive and Gram-negative pathogens
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A broad spectrum antibiotic targeting both gram positive and gram negative pathogens
Chemical Identifiers
91832-40-5 | |
395.408 | |
RTXOFQZKPXMALH-GHXIOONMSA-N | |
CHEBI:3485 | |
C=CC1=C(N2C(C(C2=O)NC(=O)C(=NO)C3=CSC(=N3)N)SC1)C(=O)O |
C14H13N5O5S2 | |
MFCD00865030 | |
6915944 | |
(6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
Specifications
Cefdinir | |
91832-40-5 | |
C14H13N5O5S2 | |
Soluble in water | |
C=CC1=C(N2C(C(C2=O)NC(=O)C(=NO)C3=CSC(=N3)N)SC1)C(=O)O | |
395.408 | |
CHEBI:3485 | |
95% | |
1 g |
Powder | |
170°C | |
MFCD00865030 | |
RTXOFQZKPXMALH-GHXIOONMSA-N | |
(6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid | |
6915944 | |
395.41 | |
White to Yellow | |
Cefdinir |
Safety and Handling
RTECSNumber : XI0367250
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only