missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Caspase-1 Inhibitor I
$154.55 - $541.96
Chemical Identifiers
CAS | 143313-51-3 |
---|---|
Molecular Formula | C23H32N4O8 |
Synonym | Ac-YVAD-CHO; Ac-Tyr-Val-Ala-Asp-CHO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ64407MCR
|
Thermo Scientific™
J64407MCR |
1 mg |
Each for $154.55
|
|
|||||
AAJ64407LB0
|
Thermo Scientific Chemicals
J64407LB0 |
5 mg |
Each for $541.96
|
|
|||||
Description
A potent, specific, and reversible inhibitor of caspases-1, -4 and -5
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
143313-51-3 | |
C23H32N4O8 | |
Ac-YVAD-CHO; Ac-Tyr-Val-Ala-Asp-CHO |
Specifications
1 mg | |
-30°C to -10°C | |
143313-51-3 | |
492.52 | |
Moisture Sensitive |
White | |
A potent, specific, and reversible inhibitor of caspases-1, -4 and -5 | |
C23H32N4O8 | |
Ac-YVAD-CHO; Ac-Tyr-Val-Ala-Asp-CHO | |
Powder |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only