missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carl Zeiss™ Microscope Dust Cover

L x W x H: 30 x 19 x 40cm

Manufacturer:  Carl Zeiss Canada Ltd 459300

Catalog No. 12-070-408


 

Specifications

Specifications

Models: standards, Axiolab™, stereomicroscopes without cameras
Dust cover K
19cm
40cm
30cm
Documents