Carl Zeiss™ C-mount Camera Adapter
Adapts specific Zeiss microscopes for use with still or video camera
Manufacturer:  Carl Zeiss™ 0000001069414
Catalog No. 12-070-354
Description
Description
Adapts microscope for use with C-mount still or video cameras; 0.63X f/60mm interface
Accessory for:
  • Stemi 2000 C /CS
  • Axiostar
  • Axioskop
  • AxioImager D1
Specifications
Specifications
Camera adapter
C mount camera
Stemi™ 2000C/CS, Axiostar™, Axioskop™, AxioImager D1
Documents