missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cargille Immersion Oil

Greatly increases light-gathering power with high-power objectives

$26.88 - $184.44

Specifications

Certifications/Compliance ISO 8036/1
Refractive Index 1.5150 ±0.0002
View More Specs

Products 11
Catalog Number Mfr. No. Density Quantity Viscosity Item Description Price Quantity  
Catalog Number Mfr. No. Density Quantity Viscosity Item Description Price Quantity  
12-370-1A View Documents Cargille
CLI16485A
0.919g/cm3 1 oz. 21000 cSt Type NVH Immersion Oil Each for $36.38
Case of 12 Each for $436.56
12-370-1B View Documents Cargille
CLI16485
0.919g/cm3 4 oz. 21000 cSt Type NVH Immersion Oil Each for $80.88
Case of 12 Each for $970.56
12-371-1A View Documents Cargille
CLI162121OZ
0.877g/cm3 1 oz. 170 cSt Type FF Immersion Oil Each for $35.34
Case of 12 Each for $424.08
12-371-1B View Documents Cargille
CLI16212C
0.877g/cm3 4 oz. 170 cSt Type FF Immersion Oil Each for $78.41
Case of 12 Each for $940.92
12-371-1C View Documents Cargille
CLI16212B
0.877g/cm3 16 oz. 170 cSt Type FF Immersion Oil Each for $184.44
Case of 12 Each for $2,213.28
12-368A View Documents Cargille
CLI16482A1
0.923g/cm3 1 oz. 150 cSt Type A Immersion Oil Each for $26.88
Case of 12 Each for $322.56
12-368B View Documents Cargille
CLI16482A16
0.923g/cm3 16 oz. 150 cSt Type A Immersion Oil Each for $124.34
Case of 12 Each for $1,492.08
12-369B View Documents Cargille
CLI16484B16
0.923g/cm3 16 oz. 1250 cSt Type B Immersion Oil Each for $124.34
Case of 12 Each for $1,492.08
12-370B View Documents Cargille
CLI16484B4
0.923g/cm3 4 oz. 1250 cSt Type B Immersion Oil Each for $63.03
12-369A View Documents Cargille
CLI16484B1
0.923g/cm3 1 oz. 1250 cSt Type B Immersion Oil Each for $26.88
Case of 12 Each for $322.56
12-370A View Documents Cargille
CLI16482A4
0.923g/cm3 4 oz. 150 cSt Type A Immersion Oil Each for $57.25
Case of 12 Each for $687.00
Description

Description

Greatly increases light-gathering power with high-power objectives

  • Formulated to give refractive index of 1.5150 ± 0.0002 for D line at 23°C (73°F)
  • Nondrying, so index will not change when exposed to air
  • Contains no polychlorinated biphenyl (PCB) compounds
  • Conforms to ISO 8036/1 specifications
  • Standard for all oil-immersion microscopy

  • Type A oil has low viscosity and low fluorescence
  • Type B is a high-viscosity oil used in biomedical microscopy
  • Type NVH is a high-viscosity oil that meets requirements for horizontal, inverted and inclined instruments and projection equipment
  • Type FF oil is virtually nonfluorescent
  • Type DF is a very low-fluorescence oil for fluorescence microscopy with very high resolution

Specifications

Specifications

ISO 8036/1
1.5150 ±0.0002