missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium Chloride, Anhydrous (Pellets, 4-20 mesh, for Desiccators), Fisher Chemical™

Supplier:  Thermo Fisher Scientific FLC614500

 View more versions of this product

Catalog No. C614-500




Description

Description

Specifications

Specifications

782°C
Solid
Beige
For desiccators
2.15g/mL
500 g
CaCl2
4 to 20 Mesh
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium dichloride
5284359
110.99
1600°C
8 to 10
Calcium Chloride Anhydrous
10043-52-4
Calcium Chloride
Pass Test
MFCD00010903
Poly Bottle
[Cl-].[Cl-].[Ca++]
110.98
CHEBI:3312

Chemical Identifiers

10043-52-4
110.98
UXVMQQNJUSDDNG-UHFFFAOYSA-L
CHEBI:3312
[Cl-].[Cl-].[Ca++]
CaCl2
MFCD00010903
5284359
calcium dichloride
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Irritating to eyes. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:1

SDS Documents

Documents

Promotions

Promotions