missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Calbiochem™ Prionex™Reagent
Used as alternative to bovine serum albumin.
Supplier: MilliporeSigma™ 529600100ML
Description
EMD Millipore Calbiochem™ Prionex™Reagent is polypeptide fraction of highly purified dermal collagen of porcine origin.
- Free from cartilage, bone plasma components
- Pure form of gelatin type A
- Used as inert protein stabilizer in many applications
- Useful as blocking agent
- Protective additive in cell culture
- Contaminants Endotoxins: ≤10 EU/mL; heavy metals: ≤20ppm
- Color: Clear, Pale Yellow
Recommended Storage
2° to 8°C
Specifications
Sterile aqueous solution pH 6.0 to 7.5 | |
100 mL |