missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Calbiochem™ Laminin Nonapeptide
Active in cell adhesion and cell-surface receptor binding and in stimulating cell migration
$188.00 - $663.00
Specifications
Purity | ≥90% by HPLC |
---|---|
Color | White to Off-white |
Content And Storage | -20°C |
Format | Lyophilized Solid |
Product Type | Laminin Nonapeptide |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
052337031MG
|
MilliporeSigma™
052337031MG |
1 mg |
Each for $188.00
|
|
|||||
052337035MG
|
MilliporeSigma™
052337035MG |
5 mg |
Each for $663.00
|
|
|||||
Specifications
≥90% by HPLC | |
-20°C | |
Laminin Nonapeptide |
White to Off-white | |
Lyophilized Solid | |
H-Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg-OH |
For Research Use Only