EMD Millipore™ Calbiochem™ H-Gly-Arg-Gly-Asp-Ser-OH
Competitively inhibits direct binding of fibroblasts to fibronectin
Manufacturer:  EMD Millipore™ 0334002725M
Catalog No. 0334002725M
Specifications
Specifications
≥97% by HPLC
White to off-white lyophilized solid
H-Gly-Arg-Gly-Asp-Ser-OH
Peptide
25mg
Supplied as an acetate salt
-20°C