missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Cadmium powder, 99.5%, Cd, CAS Number-7440-43-9, ccris 112, unii-00bh33gngh, compounds, cadmium, elemental, cadmium and compounds, 00bh33gngh, kadmium, kadmium german, colloidal, hsdb 282, 500g, (metals basis), CHEBI:22977, Silver-White

$59.42 - $1707.89

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1189109
View Documents Promotion Details
Thermo Scientific Chemicals
01189109
10 g
Each for $59.42
 
AA1189122
View Documents Promotion Details
Thermo Scientific Chemicals
01189122
100 g
Each for $390.41
 
AA1189136
View Documents Promotion Details
Thermo Scientific Chemicals
01189136
500 g
Each for $1,707.89
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-43-9
112.41
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
23973
cadmium
Cd
MFCD00010914
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
CHEBI:22977
[Cd]

Specifications

Silver-White
Cd
UN3179
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
[Cd]
112.41
CHEBI:22977
−325 Mesh
Cadmium powder
7440-43-9
MFCD00010914
10 g
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
cadmium
23973
99.5%
Solid
(metals basis)
Safety and Handling

Safety and Handling

GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.

P201-P202-P210-P240-P241-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P370+P378q-P501c

H228-H330-H341-H350-H361fd-H372

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S.

EINECSNumber : 231-152-8

RTECSNumber : EU9800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only