Bulbtronics™ Silver Oxide Battery
Manufacturer:  Bulbtronics™ 0049146
Catalog No. 14-395-896
Specifications
Specifications
40mAH
0.457 in.
Keyless-entry watches, calculator, cameras, security electronics
2.02mm
Flat
1.5V
Button or coin
11.4mm
0.08 in.
Silver Oxide
No
Silver Oxide Battery
Documents