missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution, pH 4.63 (Certified), Fisher Chemical
Supplier: Thermo Fisher Scientific SB1001
Specifications
Buffer Solution, pH 4.64 | |
64-19-7,6131-90-4,50-00-0,67-56-1,7732-18-5 | |
1.0g/cm³ | |
Liquid | |
Pass Test | |
Poly Bottle | |
Certified |
0°C | |
100°C | |
4.63 | |
1 L | |
MFCD00137248 | |
Traceable to NIST. |