Thermo Scientific™ BSA Silylation Reagent
Produce volatile, stable TMS derivatives quickly and quantitatively under mild conditions with Thermo Scientific™ BSA Silylation Reagent.
Manufacturer:  Thermo Scientific™ TS38836
 View more versions of this product
Catalog No. PI38836
Description
Description
Derivatize analytes quickly and quantitatively under mild conditions using Thermo Scientific™ BSA Silylation Reagent. This reagent reacts readily with a wide range of compounds, including alcohols, amines, carboxylic acids, phenols, steroids, biogenic amines, and alkaloids to form volatile, stable TMS derivatives for GC.
Recommended Use
Not recommended for use with carbohydrates or very low molecular weight compounds
Specifications
Specifications
2 - 8°C
Corrosive, Flammable
3 to 5%
10 x 1mL
GC
N-(Trimethylsilyl)acetamide
10 x 1mL
Documents
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.