missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BrandTech™ Short-Form Graduated Cylinders
VITLAB Short form graduated cylinders have a lower center of gravity for enhanced stability. Round based cylinders are available in polypropylene or glass-clear SAN
Supplier: BrandTech™ 645941
Description
- Choose SAN cylinders for glass clear visibility of contents
- Choose polypropylene cylinders for chemical resistance and durability
Specifications
Polypropylene | |
2.95 in. | |
20 mL | |
Round | |
11.22 in. | |
Graduated Cylinder |
±20 mL | |
78 mm | |
Autoclavable | |
Circular | |
285 mm |