BrandTech™ VACUUBRAND™ Service Kits for NT Series Chemistry Pumps and Systems
Service kits for VACUUBRAND NT series Chemistry design pumps.
$866.26 - $2243.78
Catalog Number | Mfr. No. | For Use With | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With | Price | Quantity | |||||
13-688-971
|
BrandTech™ VACUUBRAND™
20696864 |
ME 4C NT chemistry pumps and system |
Each for $866.26
|
|
|||||
13-688-976
|
BrandTech™ VACUUBRAND™
20696869 |
MZ 2C NT, MZ2C NT VARIO, PC201 NT, PC3002 VARIO |
Each for $1,052.56
|
|
|||||
13-688-974
|
BrandTech™ VACUUBRAND™
20696867 |
ME 8C NT chemistry pumps and system |
Each for $1,732.50
|
|
|||||
13-688-977
|
BrandTech™ VACUUBRAND™
20696870 |
MD 4C NT VARIO, PC3003 VARIO, PC3004 VARIO |
Each for $2,243.78
|
|
|||||
Description
- Sets include appropriate diaphragms, valves and sealing rings (if applicable)
- Wrench not included.
Specifications
Service Kit |