BrandTech™ VACUUBRAND™ BVC Shuttle for BVC Fluid Aspiration Systems
Accessory for VACUUBRAND BVC fluid aspiration systems
Supplier: BrandTech™ VACUUBRAND™ 20696880
Description
- BVC family of fluid aspiration systems deliver compact and quiet solutions for cell culture work
- Provides flexibility to accommodate decontamination protocol whether autoclaving or disinfecting in bottle
Specifications
BVC Fluid Aspiration System | |
BVC Shuttle |