Brady™ B30 Series Heavy-Duty Printer Ribbon
B30 Series “Drop & Click” print ribbons enable 5-second ribbon supply changes. No calibrating, no threading, no waste, no hassles and the ink always faces the right direction!
Supplier: Brady™ B30R6400
Description
- One-color ribbon for printing a single color onto a label
- R6400 Series Resin Formulation offers superior chemical and abrasion resistance when printed on the appropriate label material from Brady™
- Recommended for use with materials designed to resist chemicals, such as B-492 Frozen Surface Polyester and B-481 Chemical Resistant Polyester
Specifications
RoHS 2005/618/EC, MCV RoHS 2002/95/EC | |
Polyester | |
Non-sterile | |
1 Ea. |
Black | |
Conical Tube Label | |
“Drop and Click” print ribbons for 5 sec. |