Bradley™ Replacement Liners for Gravity-Fed Eyewash
Liner for use with Bradley portable eyewash unit.
Supplier: Bradley™ 2691379
Description
- For insertion into protective cap on tank bottom.
- Liners are pierced when tank is mounted on pedestal and must be replaced each time water is refilled.
Specifications
White | |
Portable Eyewash | |
Paper | |
Liner |