missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BOC-L-Serine hydrate, 98%, Thermo Scientific Chemicals
$310.40 - $310.40
Chemical Identifiers
CAS | 3262-72-4 |
---|---|
Molecular Formula | C8H15NO5 |
Molecular Weight (g/mol) | 205.21 |
MDL Number | MFCD00037243 |
InChI Key | FHOAKXBXYSJBGX-MLHKIVSYNA-N |
Synonym | boc-ser-oh, n-boc-l-serine, boc-l-serine, n-tert-butoxycarbonyl-l-serine, n-boc-serine, s-n-boc-serine, n-tert-butoxycarbonyl serine, s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, n-t-butoxycarbonyl-l-serine, boc-l-ser |
PubChem CID | 98766 |
SMILES | CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC275660250
|
Thermo Scientific Chemicals
275660250 |
25 g | Glass Bottle |
Each for $310.40
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3262-72-4 | |
205.21 | |
FHOAKXBXYSJBGX-MLHKIVSYNA-N | |
98766 |
C8H15NO5 | |
MFCD00037243 | |
boc-ser-oh, n-boc-l-serine, boc-l-serine, n-tert-butoxycarbonyl-l-serine, n-boc-serine, s-n-boc-serine, n-tert-butoxycarbonyl serine, s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, n-t-butoxycarbonyl-l-serine, boc-l-ser | |
CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O |
Specifications
84.0°C to 89.0°C | |
White | |
C8H15NO5 | |
HOCH2CH(COOH)NHCOOC(CH3)3· | |
Glass Bottle | |
FHOAKXBXYSJBGX-MLHKIVSYNA-N | |
(2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
205.21 | |
205.21 | |
Technical | |
25 g | |
Powder |
3262-72-4 | |
98% | |
Authentic | |
MFCD00037243 | |
boc-ser-oh, n-boc-l-serine, boc-l-serine, n-tert-butoxycarbonyl-l-serine, n-boc-serine, s-n-boc-serine, n-tert-butoxycarbonyl serine, s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, n-t-butoxycarbonyl-l-serine, boc-l-ser | |
CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O | |
− 8.10 | |
98766 | |
98% | |
7.5 to 8.0% (K.F.) | |
186°C | |
BOC-L-Serine hydrate |
RUO – Research Use Only