Learn More
BOC-D-Phenylalanine, 99+%, Thermo Scientific Chemicals
$131.78 - $131.78
Chemical Identifiers
CAS | 18942-49-9 |
---|---|
Molecular Formula | C14H19NO4 |
Molecular Weight (g/mol) | 265.31 |
MDL Number | MFCD00063149 |
InChI Key | ZYJPUMXJBDHSIF-LLVKDONJSA-N |
Synonym | boc-d-phenylalanine, boc-d-phe-oh, n-boc-d-phenylalanine, n-tert-butoxycarbonyl-d-phenylalanine, boc-d-phe, d-phenylalanine, n-1,1-dimethylethoxy carbonyl, tert-butoxycarbonyl-d-phenylalanine, n-alpha-t-butyloxycarbonyl-d-phenylalanine, 2r-2-tert-butoxy carbonylamino-3-phenylpropanoic acid, 2r-2-tert-butoxycarbonyl amino-3-phenylpropanoic acid |
PubChem CID | 637610 |
SMILES | CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC275740050
|
Thermo Scientific Chemicals
275740050 |
5 g | Glass Bottle |
Each for $131.78
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18942-49-9 | |
265.31 | |
ZYJPUMXJBDHSIF-LLVKDONJSA-N | |
637610 |
C14H19NO4 | |
MFCD00063149 | |
boc-d-phenylalanine, boc-d-phe-oh, n-boc-d-phenylalanine, n-tert-butoxycarbonyl-d-phenylalanine, boc-d-phe, d-phenylalanine, n-1,1-dimethylethoxy carbonyl, tert-butoxycarbonyl-d-phenylalanine, n-alpha-t-butyloxycarbonyl-d-phenylalanine, 2r-2-tert-butoxy carbonylamino-3-phenylpropanoic acid, 2r-2-tert-butoxycarbonyl amino-3-phenylpropanoic acid | |
CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C(O)=O |
Specifications
78.0°C to 87.0°C | |
White | |
C14H19NO4 | |
C6H5CH2CH(COOH)NHCOOC(CH3)3 | |
Glass Bottle | |
Solubility in water: insoluble. Other solubilities: soluble in organic solvents | |
CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C(O)=O | |
− 25.00 | |
637610 | |
≥99% | |
5 g | |
BOC-D-Phenylalanine |
18942-49-9 | |
99+% | |
Authentic | |
MFCD00063149 | |
boc-d-phenylalanine, boc-d-phe-oh, n-boc-d-phenylalanine, n-tert-butoxycarbonyl-d-phenylalanine, boc-d-phe, d-phenylalanine, n-1,1-dimethylethoxy carbonyl, tert-butoxycarbonyl-d-phenylalanine, n-alpha-t-butyloxycarbonyl-d-phenylalanine, 2r-2-tert-butoxy carbonylamino-3-phenylpropanoic acid, 2r-2-tert-butoxycarbonyl amino-3-phenylpropanoic acid | |
ZYJPUMXJBDHSIF-LLVKDONJSA-N | |
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid | |
265.31 | |
265.3 | |
0.5% max. (K.F.) | |
Fine Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only