missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Innovating Science™ Blood Typing Kits with Simulated Blood
Determine the ABO or ABO/Rh blood type of four unknown samples.
$77.82 - $139.56
Specifications
Certifications/Compliance | 49 CFR 173.4 |
---|---|
Color | White |
Grade | 7 to 14 |
Age | 13+ years |
Quantity | 1 |
Includes:
- Four simulated blood samples
- One bottle each of simulated anti-A and anti-B serum
- 40 blood typing trays
- ABO/Rh kit also includes one bottle of simulated anti-Rh serum
Catalog Number | Mfr. No. | For Use With (Application) | Includes | Product Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Includes | Product Type | Price | Quantity | |||
S96792
|
Innovating Science™
IS3101 |
For simulated abo/rh blood typing | 4 simulated blood samples, 1 bottle simulated anti-a serum, 1 bottle simulated anti-b serum, 1bottle simulated anti-rh serum, 40 blood typing trays, Teacher's manual and student study guide copymasters | Blood Type Kit |
Each for $139.56
|
|
|||
S96792R
|
Innovating Science™
IS3101REF |
For simulated abo/rh blood typing | Instructions, msds packet | Innovating Science™ Refill |
Each for $77.82
|
|
|||
Description
Use hands-on activities to understand the nature and importance of antigen-antibody reactions.
- Provide accurate simulation of the blood typing procedure
- No toothpicks, stirring or waiting for results
- Kit contains materials for 10 groups
Kits use simulated blood only.
Order Info
Kit refills do not include blood typing trays.
Specifications
49 CFR 173.4 | |
7 to 14 | |
1 |
White | |
13+ years | |
10 Lab Groups |