missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Biocontainment Lid for Thermo Scientific™ Sorvall™ Microplate Rotor 75-002-010

Optional biocontainment-approved lids offered in pairs

Manufacturer:  Thermo Scientific™ 75002011

Catalog No. 75002011



Description

Description

Accessory for Sorvall™ MP 3300 Microplate Rotor

For use with Sorvall microplate rotor 75-002-010. Sold in pairs.

Specifications

Specifications

MP 3300 Microplate Rotor accessory
75-002-010 Microplate rotor
Documents