Learn More
Agilent BioTek Synergy LX Multi-Mode Reader
The BioTek Synergy LX multi-mode reader automates many common microplate assays. For Research Use Only. Not for use in diagnostic procedures.
Specifications
Absorbance Range | 200 to 900 nm |
---|---|
Accuracy | Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Bandwidth | ≤5 nm |
Depth (Metric) | 38.1 cm |
Catalog Number | Mfr. No. | Model | Includes | Height (English) | Height (Metric) | Price | Qty | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Includes | Height (English) | Height (Metric) | Price | Qty | ||
BTSLXA
|
Agilent BioTek
SLXASN |
SLXA | Absorbance | 12 in. | 30.5 cm |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTSLXATS
|
Agilent BioTek
SLXATSSN |
SLXATS | Absorbance; Touchscreen | 15 in. | 38.1 cm |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTSLXF
|
Agilent BioTek
SLXFSN |
SLXF | Fluorescence; Luminescence | 12 in. | 30.5 cm |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTSLXFTS
|
Agilent BioTek
SLXFTSSN |
SLXFTS | Fluorescence; Luminescence; Touchscreen | 15 in. | 38.1 cm |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTSLXFA
|
Agilent BioTek
SLXFASN |
SLXFA | Absorbance; Fluorescence; Luminescence | 12 in. | 30.5 cm |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTSLXFATS
|
Agilent BioTek
SLXFATSSN |
SLXFATS | Absorbance; Fluorescence; Luminescence; Touchscreen | 15 in. | 38.1 cm |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The BioTek Synergy LX multi-mode reader automates many common microplate assays. The high-quality optic design ensures excellent data in absorbance, fluorescence, and luminescence detection modes. This affordable system is ideal for many common assays including nucleic acid and protein quantification, ELISA, BCA, Bradford, and cell viability
The data display, output to a USB flash drive, printer, or Gen5 software make Synergy LX a versatile assay workstation. The large touch screen user interface and onboard software simplify programming and save time in operation of microplate and microvolume assays.
Warranty and Services
12 Months
Specifications
200 to 900 nm | |
CE and TUV marked. RoHS compliant. | |
38.1 cm | |
24 sec. (96 wells), 76 sec. (384 wells) | |
Monochromator Bandpass | |
50/60 Hz | |
Multi-mode Microplate Reader | |
0.001 OD | |
2 | |
1 year | |
27 lb. | |
15 in. | |
Photodiode, PMT | |
End Point, Kinetic, Area Scanning, Absorbance Spectral Scanning |
Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD | |
≤5 nm | |
15 in. | |
Linear, Orbital, Double Orbital | |
Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability | |
<1% from 0 to 2.5 OD | |
24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.) | |
Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP | |
English | |
100/240 V | |
12.3 kg | |
38.1 cm | |
Xenon Flash |