missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Bio-Cane™ Cane and Canister Systems

Store cells, tissue and biologicals in a cost-effective manner with Thermo Scientific™ Bio-Cane™ Canister and Cane Systems.

$3,642.04 - $7,864.92

Specifications

Certifications/Compliance CE
View More Specs

Products 4
Catalog Number Mfr. No. Capacity (Metric) No. of Canes/Vials per Cane Static Holding Time Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) No. of Canes/Vials per Cane Static Holding Time Price Quantity  
11-675-91 Accessories View Documents Thermo Scientific™
CK509X2
20.5L/day 5 /150 227 days Each for $3,642.04
11-675-92 Accessories View Documents Thermo Scientific™
CK509X3
34.8L/day 20 /630 193 days Each for $4,340.63
11-675-93 Accessories View Documents Thermo Scientific™
CK509X4
47.4L/day 42 /1320 121 days Each for $5,516.04
13-762-351 View Documents Thermo Scientific™
PSICK509X6
73L/day 52 /2400 121 days Each for $7,864.92
Description

Description

Store cells, tissues and biologicals in an economical and biological manner with Thermo Scientific™ Bio-Cane™ Cane and Canister Systems. Available in four sizes, the system can safely hold samples for up to seven months without replenishing LN2.

Features:
  • Six stainless-steel canisters to accommodate vials (eight with Bio-Cane 70)
  • Canisters are numbered and color-coded to simplify sample identification
  • Durable aluminum construction and vacuum insulation
  • Narrow-mouth design minimizes LN2 evaporation
  • Secure locking clasp

Includes:

Six canisters

Compatible with:

  • Cart, Wheeled (BioCane 20, 34, 47: AY509X9) (BioCane 73: AY509X1)
  • Thermo Scientific™ Cryosleeve™, 100/case, (AY509X21)
  • Thermo Scientific™ Cryocane™, 1.2mL (4000211)
  • Rack, Square, for BioCane 47, includes five boxes with 40 dividers (195555)
  • Vials, Cryogenic, 1.2mL, case of 500 (AY509X13)
  • Vials, Cryogenic, 2.0mL, case of 500 (AY509X12)
  • Vials, 1mL, case of 500 (AY509X32)
  • Vials, 1.5mL, case of 500 (AY509X33)
  • Warranty varies by country, please contact us for details
Specifications

Specifications

CE