missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Spindrive™ Orbital Shaker Platform

Turns a standard magnetic stirrer into an orbital shaking platform saving the expense and space requirements of additional lab equipment.

Manufacturer:  Bel-Art™ F370410000

Catalog No. 14-511-76


 

Description

Description

Perfect for dishes, flasks, and trays, it is also ideal for Western Blot processing.

  • Shake and mix; rotating orbit diameter of 21.7mm (7/8 in.)
  • Shaker adjustment is controlled by setting the magnetic stirrer to the desired speed
  • Platform protects the operating mechanism from spills
  • Non-skid surface prevents containers from sliding
  • Base incorporates built-in hand holds for lifting and carrying
  • Supports up to 6.8 kg (15 lb.)
  • Dimensions: 8.9 x 8.9 x 2.1 in.
  • Weight: 2.2 lb.

U.S. Patent 5,409,312.

Specifications

Specifications

8.9 in.
2.25 lb.
2.1 in.
Orbital platform shaker
22.5cm
22.5cm
22.5cm
1kg
5.3cm
8.9 in.
8.9 x 8.9 x 2.1 in. (22.5 x 22.5 x 5.3cm)
8.9 in.
Documents