missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Spinbar™ Magnetic Retriever

Reaches down drains for lost bars

Manufacturer:  Bel-Art™ F377730000

Catalog No. 22-269746




Description

Description

  • 0.38 in. (9.5mm) dia. polyethylene pick-up rod
  • With powerful Alnico magnet sealed in one end
  • Handy hang-up grip on other end
  • Offers convenient stir bar retrieval for bars up to 1 in. long

Specifications

Specifications

Offers convenient stir bar retrieval for bars up to 1 in. long
High-strength alnico sealed-in magnet, hang-up loop on one end, 0.95 cm polyethylene pick-up rod
Spinbar™ Magnetic Retriever
PE (Polyethylene)
Stirring bars
18 in.
45.7cm
Documents