Bel-Art™ Pipette Pump™ Pipetters
Provides safe, accurate and trouble-free pipetting. Bel Art™ SP Scienceware™ Pipette Pump™ Pipetters utilize an easy-to-use one-handed operation.
$6.77 - $80.12
Specifications
Type Pipette
For Use With (Equipment) Pipets
Resistance Acid and Alkali
View More Specs
Products 4
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity  
13-683B
View Documents Promotion Details
Bel-Art™
F378970000
Blue 2 mL
Each for $54.88
Case of 12 Each for $658.56
13-683A
View Documents Promotion Details
Bel-Art™
F378960000
Yellow 0.2 mL
Each for $6.77
Case of 12 Each for $81.24
13-683C
View Documents Promotion Details
Bel-Art™
378980000
Green 10 mL
Each for $63.12
Case of 12 Each for $757.44
13-683D
View Documents Promotion Details
Bel-Art™
378990000
Red 25 mL
Each for $80.12
Case of 12 Each for $961.44
Description
Description
Provides safe, accurate and trouble-free pipetting. Bel Art™ SP Scienceware™ Pipette Pump™ Pipetters utilize an easy-to-use one-handed operation.
  • Pipettes fit smoothly into its flexible tapered chuck
  • Thumbwheel on side is rotated for precision aspirating or dispensing
  • Plunger may be pressed for quick emptying
  • Resistant to acids and alkalis
  • Disassemble all parts for cleaning and before autoclaving at 121ºC (250ºF) for 20 minutes; chuck should not be autoclaved
  • Replacement Silicone Rubber Chuck and Washers are available 
Specifications
Specifications
Pipette
Pipets
Acid and Alkali
SDS
missing translation for 'documents'
missing translation for 'documents'