Bel-Art™ SP Scienceware™ Piezoelectric Gas Lighter
Needs no recharging
Supplier:  Bel-Art™ H150560000
Catalog No. 12-002
Description
Description
Needs no recharging
  • Compact, self-contained electric generator lights Bunsen burners, gas stoves, ovens, propane torches, and oxyacetylene torches
  • Just hold in gas stream and press button
  • Functions without flints, fuel, batteries, or wires
  • Comfortable plastic pistol grip provides excellent control in aiming aluminum barrel
Order Info
Dimensions: 10 3/8L x 1W x 5 in.H (26 x 2.5 x 13cm).
Specifications
Specifications
Piezo Electric Gas Lighter
26.5 cm
Comfortable plastic pistol grip provides excellent control
10.438 in.
Plastic