missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Long Rigid Scraper

Flat scraper has a straight, chisel-type, scraping edge which may be resharpened

Manufacturer:  Bel-Art™ H368200000

Catalog No. 22-261589



Description

Description

  • Made from glass-reinforced polyimide nylon
  • Blade length: 14cm (5.5 in.)
  • Highest temperature: 180°F (82°C)

Specifications

Specifications

5.5 in.
2.5 in.
11.2 in.
Rigid Scraper
14cm
6.3cm
28.5cm
Glass-reinforced Polyimide Nylon
Documents