Bel-Art™ SP Scienceware™ Complete Fittings Assortment
Reduces time spent searching for the right tubing connector or stopper
Supplier: Bel-Art™ F196250000
Description
This generous assortment of 72 plastic fittings has everything needed for routine laboratory set-ups.
- Clear, hinge-top, polystyrene storage box makes it easy to find what you need
- Includes commonly used sizes of connector and stopper styles
- FDA grade, autoclavable polypropylene connectors: 15 Straight Tubing Connectors, 11 Stepped Tubing Connectors, 6 “T” Shaped Tubing Connectors, 10 Wye (Y) Tubing Connectors
- Polyethylene connectors: 3 pair Quick Disconnects
- Polyethylene stoppers: 24 Cup-Type
Specifications
Fittings Assortment | |
Assortment of 72 plastic fittings |