missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Bent Handle Scoop
Features an ergonomic, curved handle design to elevate the hand from the sample, reducing risk of contamination
$105.38 - $105.38
Specifications
Product Type | Scoop |
---|---|
Material | Polystyrene |
Bottom Type | Flat |
Handle Type | Bent and curved |
Disposable | Yes |
Catalog Number | Mfr. No. | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity | ||||
14-241-202
|
Bel-Art™
H369022006 |
60 mL | 7.62 x 1.62 x 1.12cm (19.4 x 4.1 x 2.9 cm) |
Pack of 10 Each for $105.38
Case of 10 Pack for $1,053.80
|
|
||||
Description
- Curved handle helps prevent incidental contact with sample source material and exposure to hazardous substances
- Flat bottom keeps scoop upright when placed on a flat surface; can be used as a weigh boat
- Made from high-impact, lightweight, FDA grade polystyrene
- Sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Sterilization
- Individually packaged and ready for one time use and disposal
Specifications
Scoop | |
Flat | |
Yes | |
FDA | |
7.62 in. |
Polystyrene | |
Bent and curved | |
Sterile | |
Design to elevate the hand from the sample, reducing risk of contamination, prevent incidental contact with sample source material and exposure to hazardous substances | |
19.4 cm |