missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Tuberculin Syringe

Manufacturer:  BD B305620

Catalog No. 14-829-9




Description

Description

  • Single-use
  • Permanently attached needle
  • Single-use
  • Permanently attached, stainless-steel PrecisionGlide™ needle
  • Point has regular bevel

Order Info

Packaged individually in easy peel blister package.

Specifications

Specifications

Gray
0.5mL
Permanently attached needle
1.3cm
Bevel
Tuberculin Syringe
27 ga.
Yes
0.5 in.
Stainless steel
Sterile
Documents
Accessories