BD Disposable Syringes with Luer-Lok™ Tips
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
Supplier: BD 309646
Description
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
- Sterile syringe for single use
- Without needle
- Clear barrel (polycarbonate or polypropylene) with bold scale markings
- Tapered plunger rod for ease of aspiration
- Positive plunger rod stop
- Added BD Luer-Lok™ thread for increased secure connection
- DNR (Latex) free
- Shelf life: 1825 days
Specifications
Polycarbonate | |
Syringe | |
5 mL | |
Sterile |
Yes | |
Yes | |
Luer Lock |