missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Vacutainer™ Slip-tip Syringe

Nonsterile slip-tip syringe

Manufacturer:  BD B301036

Catalog No. 14-820-12



Description

Description

  • Volume: 60mL (2 oz.)
  • Bulk
  • Shelf-life: 1825 days

Specifications

Specifications

60mL
Non-sterile
2.02 oz.
Plastic
Syringe
Documents