missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Clay Adams™ TRIAC™ Combination Centrifuge

Three-speed centrifuge for separation of serum from blood, urine sedimentations and microhematocrit determinations

Manufacturer:  BD B420200

Catalog No. 0510080


 

Includes: Complete with microhematocrit rotor and four trunnion rings, a vial of microhematocrit tubes, a tray of sealing clay, and eight urine tubes

Description

Description

Operating Features

  • Capacity: 12 capillary tubes
  • Maximum speed/force: 10,400rpm/12,600 x g
  • 12-place, direct-reading microhematocrit rotor accommodates four trunnion rings for 8-place blood and urine separation capacity
  • Unit encompasses three runtime/speed combinations, plus an electric brake and timer
  • In blood mode, unit provides ample power to handle serum separation devices
Safety Features
  • Rugged molded housing with steel-reinforced bowl gives extra protection
  • Microhematocrit rotor interlock switch and lid safety switch prevent starting the centrifuge with cover open
  • Centrifuge automatically shuts off if opened during operation

Specifications

Specifications

Electronic Brake
Yes, Electric
10,400rpm
120V 60Hz
53.3cm
45.7cm
18.25 in.
12,000 to 12,999 x G
No
26 lb. (11.8kg)
12,600 x G
No
21 in.
18 in.
Rugged molded housing with steel-reinforced bowl gives extra protection; Microhematocrit rotor interlock switch and lid safety switch prevent starting the centrifuge with cover open; Centrifuge automatically shuts off if opened during operation
46.3cm
10,000 to 10,999rpm
Documents