Learn More
Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables.
$480.30 - $4145.97
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
09-050-284
|
Thermo Scientific™
D50280 |
R/O and distilled feed; Ultralow organics, type I water |
Each for $3,952.18
|
|
|||||
09-050-283
|
Thermo Scientific™
D50281 |
Deionized water feed ; Ultralow organics, type I water |
Each for $3,952.18
|
|
|||||
09-035-13
|
Thermo Scientific™
D50282 |
R/O and distilled feed; Low organics, type I water |
Each for $2,392.69
|
|
|||||
09-050-282
|
Thermo Scientific™
D50283 |
Deionized water feed; Low organics, type I water |
Each for $4,145.97
|
|
|||||
09-035-16
|
Thermo Scientific™
E896X5 |
Calibration Module |
Each for $480.30
|
|
|||||
Description
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
- Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter