missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Baby Bears™ Balance Set

Explore measurement with Baby Bears!

Manufacturer:  Fisher Scientific LER0779

Catalog No. S90834


Alert:

CHOKING HAZARD —Small parts. Not for children under 3 yrs.


Includes: Balance plus 102 bears in six rainbow colors (17 of each color).

Description

Description

  • Clear balance buckets remove easily for emptying
  • Cover buckets with lids for use as measurement platforms
  • Buckets can also be used to measure liquids
  • Balance features a built-in storage drawer and measures 16.5 × 6.5 × 6 in.
  • Each Baby Bear measures 1 in. and weighs 4 grams

Order Info

Grades PK+.

Specifications

Specifications

Baby Bear™ balance set
4g
Balance plus 102 bears in six rainbow colors (17 of each color)
Documents