Axygen™ Aerosol Filter Tips, Sterile
Prevent cross-contamination with robotic barrier tips. Axygen™ Aerosol Filter Tips, Sterile are e-beam irradiated to prevent product degradation and are free of leachable additives. Available in a wide variety of tip styles.
$243.57 - $1815.35
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Color | Clear |
Clean Claims | DNase, RNase, and Pyrogen Free |
Material | Polypropylene |
Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Tip Style | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Tip Style | Price | Quantity | |||
14-222-219
|
Axygen™
MTXF1250CRS |
15 to 1250 μL | Sterile | Matrix-style |
Pack of 1152 Each for $750.96
Case of 3 Pack for $2,252.88
|
|
|||
14-222-763
|
Axygen™
TF100 |
1 to 100 μL | Sterile | Universal |
Pack of 1000 Each for $243.57
Case of 10 Pack for $2,435.70
|
|
|||
14-222-765
|
Axygen™
TF1000LRS |
100 to 1000 μL | Sterile | Maxymum Recovery |
Pack of 1000 Each for $337.57
Case of 5 Pack for $1,687.85
|
|
|||
14-222-770
|
Axygen™
TF100RS |
1 to 100 μL | Sterile | Universal |
Pack of 960 Each for $320.96
Case of 5 Pack for $1,604.80
|
|
|||
14-222-766
|
Axygen™
TF1000RS |
100 to 1000 μL | Sterile | Universal |
Pack of 1000 Each for $315.48
Case of 5 Pack for $1,577.40
|
|
|||
14-222-769
|
Axygen™
TF100LRS |
1 to 100 μL | Sterile | Maxymum Recovery |
Pack of 960 Each for $358.17
Case of 5 Pack for $1,790.85
|
|
|||
14-222-772
|
Axygen™
TF1200CRS |
100 to 1100 μL | Sterile | Universal |
Case of 3456 Each for $1,468.43
|
|
|||
50-400-962
|
Axygen™
TF150RS |
1 to 150 μL | Sterile | Universal |
Case of 4800 Each for $1,684.57
|
|
|||
14-222-777
|
Axygen™
TF200LRS |
1 to 200 μL | Sterile | Maxymum Recovery |
Pack of 960 Each for $351.78
Case of 5 Pack for $1,758.90
|
|
|||
14-222-778
|
Axygen™
TF200RS |
1 to 200 μL | Sterile | Universal |
Pack of 960 Each for $315.48
Case of 5 Pack for $1,577.40
|
|
|||
14-222-781
|
Axygen™
TF20LRS |
0.5 to 20 μL | Sterile | Maxymum Recovery |
Pack of 960 Each for $351.78
Case of 5 Pack for $1,758.90
|
|
|||
14-222-782
|
Axygen™
TF20RS |
0.5 to 20 μL | Sterile | Universal |
Pack of 960 Each for $315.48
Case of 5 Pack for $1,577.40
|
|
|||
14-222-787
|
Axygen™
TF350LRS |
1 to 300 μL | Sterile | Maxymum Recovery |
Pack of 960 Each for $351.78
Case of 5 Pack for $1,758.90
|
|
|||
14-222-788
|
Axygen™
TF350RS |
1 to 300 μL | Sterile | Universal |
Pack of 960 Each for $321.22
Case of 5 Pack for $1,606.10
|
|
|||
14-222-794
|
Axygen™
TF420LRS |
0.5 to 20 μL | Sterile | Maxymum Recovery, Ultra Micro |
Pack of 960 Each for $376.74
Case of 5 Pack for $1,883.70
|
|
|||
14-222-795
|
Axygen™
TF420RS |
0.5 to 20 μL | Sterile | Ultra-micro |
Case of 4800 Each for $1,683.74
|
|
|||
14-222-799
|
Axygen™
TF50LRS |
0.5 to 50 μL | Sterile | Maxymum Recovery |
Case of 960 Each for $1,815.35
|
|
|||
14-222-800
|
Axygen™
TF50RS |
0.5 to 50 μL | Sterile | Universal |
Case of 4800 Each for $1,617.17
|
|
|||
Description
- Technologically advanced filter barrier pipette tips
- Ideal for DNA amplification and microbiological applications where trace contamination can invalidate results
- Filter allows free passage of air while preventing cross-contamination by blocking the passage of aerosols or liquids
- Nonpyrogenic, clear polyethylene filters are free from DNA inhibitors and leachable additives
- Every filter is inserted into the tip robotically to ensure freedom from nuclease contamination
- Packaged in sealed-bottom racks with lids to prevent contamination
- Wide variety of tip styles available in a range of packaging quantities:
- Maxymum Recovery™ technology provides excellent accuracy and prevents wastage of precious samples
- Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
Order Info
Available in bulk or racked formats.
Certifications
Certified RNase-, DNase- and Pyrogen free
Specifications
Filtered | |
Clear | |
Polypropylene |
Sterile | |
DNase, RNase, and Pyrogen Free |