Aushon CiraScan™ Imaging and Analysis System DFS Item
Complete system includes an improved CCD imager with custom optics designed specifically for quantitative chemiluminescent protein arrays
Manufacturer:  Aushon Biosystems Inc 000026FA
Catalog No. 000026FA
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
User-Friendly System
  • Includes imager integrated with embedded Mac mini computer and iPad user interface
  • integrated touch screen interface for image capture and storage
  • Fixed-focus settings and automatic calculation of optimal exposure times
  • Generates 16-bit images in under two minutes
Improved Productivity
  • Custom, high-resolution camera/lens improves image quality and consistency of results
  • Analyze images at the lab bench or desk, freeing imager for other activities
Superior Results
  • Cirasoft™ software provides fast, accurate, reproducible analysis
  • Customized array software automatically locates each spot in the image
  • Measures density values and calculates pg/mL values for unknown samples from standard curves
Documents