missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Arsenic III Speciation Standard (Assurance™ Grade/For ICP-MS), SPEX CertiPrep™
Supplier: SPEX CertiPrep Group LLC SPECAS3M
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
As(+3) | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Arsenic III Speciation Standard | |
Triple-leached LDPE bottle | |
0.1,2.0,97.9 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
As | |
RQNWIZPPADIBDY-UHFFFAOYSA-N | |
arsenic | |
5359596 | |
Assurance |
7440-38-2,7647-01-0,7732-18-5 | |
30 mL | |
0.1,2.0,97.9 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00085309 | |
[As] | |
74.92 | |
CHEBI:27563 | |
2% HCl |
Chemical Identifiers
7440-38-2 | |
74.92 | |
RQNWIZPPADIBDY-UHFFFAOYSA-N | |
CHEBI:27563 | |
[As] |
As | |
MFCD00085309 | |
5359596 | |
arsenic |
Safety and Handling
Causes skin irritation, serious eye irritation
DOTInformation : Nonhazardous
Recommended Storage : Ambient