missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thyroid Stimulating Hormone, clone: Fc1003, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Pierce Biotechnology Inc MA514738

Catalog No. PIMA514738



Description

Description

The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family.

MA5-14738 targets Thyroid Stimulating Hormone in ELISA, IP, and RIA applications and shows reactivity with Human samples.The MA5-14738 immunogen is human Thyroid Stimulating Hormone.MA5-14738 detects Thyroid Stimulating Hormone which has a predicted molecular weight of approximately 10 kDa.Product MA514738 is a smaller package size of MIT0406 (formerly sold as a Seradyn product).
Specifications

Specifications

Thyroid Stimulating Hormone
Monoclonal
1mg/mL
0.01M potassium phosphate with 0.135M NaCl, 1mg/mL BSA, 2.5mM KCl and 0.05% sodium azide
P01215
FSH-alpha, LSH-alpha, TSH-alpha, anterior pituitary glycoprotein hormones common subunit alpha, choriogonadotropin alpha chain, chorionic gonadotrophin subunit alpha, chorionic gonadotropin, alpha polypeptide, follicle-stimulating hormone alpha chain, follicle-stimulating hormone alpha subunit, follitropin alpha chain, glycoprotein hormones alpha chain, luteinizing hormone alpha chain, lutropin alpha chain, thyroid-stimulating hormone alpha chain, thyrotropin alpha chain
Murine
IgG1
100μg
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
1081
ELISA, Immunoprecipitation, Radioimmune Assays
Fc1003
Unconjugated
CGA
Liquid
CGA
Human Thyroid Stimulating Hormone
Protein A
RUO
Antibody
Monoclonal
Human
Documents