missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MAP2 Mouse anti-Human, Mouse, Rat, Rabbit, Clone: 5F9, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05346
Description
Detect MAP2 using this Anti-MAP2 Antibody, clone 5F9 validated for use in western blot, immunohistochemistry.Specifications
MAP2 | |
Monoclonal | |
Unconjugated | |
NM_031847 | |
Mouse | |
Protein G chromatography | |
Primary | |
−20°C for 24 months. | |
IgG1 |
Immunohistochemistry, Western Blot | |
5F9 | |
200μg of protein G purified mouse IgG1 in 200μL of storage buffer (0.02Mphosphate buffer, pH 7.6, 0.25MNaCl, and 0.1% sodium azide. Frozen at −20°C. | |
MAP2, MAP2A, MAP2B | |
Purified rat brain microtubules MAP2 | |
200 μg | |
Human, Mouse, Rabbit, Rat | |
Purified |
For Research Use Only