missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ansell AlphaTec™ Solvex™ 37-145 Nitrile Gloves
Solvex™ 11 mil nitrile gloves are tactile and durable and can be used in a diverse range of applications including automotive, machinery/equipment and chemical.
$.64 - $7.79
Specifications
ANSI Abrasion Rating | ANSI 5 |
---|---|
Certifications/Compliance | FDA 21 CFR 177.2600 |
Chlorinated | Yes |
Color | Green |
Cut Resistance Rating | ANSI A4 |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
11-391-136A
|
Ansell
3714511060 |
6 |
Pack of 12 Pair for $7.79
Case of 12 Pack for $93.48
|
|
|||||
11-391-136B
|
Ansell
3714511070 |
7 |
Pair for $0.64
Case of 144 Pair for $92.16
|
|
|||||
11-391-136C
|
Ansell
3714511080 |
8 |
Pair for $2.28
Case of 144 Pair for $272.16
|
|
|||||
11-391-136D
|
Ansell
3714511090 |
9 |
Pair for $2.28
Case of 144 Pair for $272.16
|
|
|||||
11-391-136E
|
Ansell
3714511100 |
10 |
Pair for $2.28
Case of 144 Pair for $272.16
|
|
|||||
11-391-136F
|
Ansell
3714511110 |
11 |
Pair for $2.03
Case of 144 Pair for $241.92
|
|
|||||
Description
Solvex™ 11 mil nitrile gloves are tactile and durable and can be used in a diverse range of applications including automotive, machinery/equipment and chemical.
- Nitrile construction protects against abrasions, cuts, snags and punctures
- Curved finger design and contoured palm help reduce fatigue
- Full cut for easy donning and doffing
- Dark Green
- No inner cotton flocking, reducing risk of lint contamination
- Sandpatch finish provides smooth surface
- Fully reusable, comfortable
- Complies with FDA food handling requirements
Specifications
ANSI 5 | |
Yes | |
ANSI A4 | |
Nitrile | |
0.27 mm | |
No | |
JKNOPT | |
Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work | |
Sandpatch | |
13 in. |
FDA 21 CFR 177.2600 | |
Green | |
Yes | |
11 mil | |
EN 388:4001, EN 388:3001X | |
Type A | |
Bacteria, Fungi only | |
Gauntlet | |
Unlined | |
33 cm |