missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

eBioscience™ Annexin V-FITC Apoptosis Detection Kit

Annexin V staining, paired with 7-AAD or PI is widely used to identify apoptotic stages by flow cytometry.

$149.00 - $524.00

Specifications

For Use With (Application) Flow Cytometric Analysis
Synonym Lipocortin V, Endonexin II, Calphobindin I
Assay > 98%
Conjugate FITC
Formula Weight 35.8 kd
View More Specs

Includes: Annexin V-FITC protein
Binding Buffer (4x)
Propidium Iodide (20 ug/mL)

Products 3 Description

Description

  • Blue Laser
  • Excitation at 488nm
  • Emission at 520nm
Specifications

Specifications

Flow Cytometric Analysis
> 98%
35.8 kd
Lipocortin V, Endonexin II, Calphobindin I
FITC

For Research Use Only