Learn More
Anisole, 99%, Thermo Scientific Chemicals
$88.90 - $250.60
Chemical Identifiers
CAS | 100-66-3 |
---|---|
Molecular Formula | C7H8O |
Molecular Weight (g/mol) | 108.14 |
MDL Number | MFCD00008354 |
InChI Key | RDOXTESZEPMUJZ-UHFFFAOYSA-N |
Synonym | methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene |
PubChem CID | 7519 |
ChEBI | CHEBI:16579 |
IUPAC Name | anisole |
SMILES | COC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1299736
|
Thermo Scientific Chemicals
A1299736 |
500 g |
Each for $88.90
|
|
|||||
AAA129970E
|
Thermo Scientific Chemicals
A129970E |
2500 g |
Each for $250.60
|
|
|||||
Description
Anisole is widely used as a solvent for the synthesis of various organic compounds, anethole, nonylphenol isomer 4-(3',6'-dimethyl-3-heptyl)phenol, perfumes, insect pheromones and pharmaceuticals. It finds application in the preparation of inorganic complexes and materials such as tin-core/tin oxide nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAnisole is widely used as a solvent for the synthesis of various organic compounds, anethole, nonylphenol isomer 4-(3′,6′-dimethyl-3-heptyl)phenol, perfumes, insect pheromones and pharmaceuticals. It finds application in the preparation of inorganic complexes and materials such as tin-core/tin oxide nanoparticles.
Solubility
Miscible with methanol and acetone. Immiscible with water.
Notes
Keep away from sources of ignition. Incompatible with strong oxidizing agents.
Chemical Identifiers
100-66-3 | |
108.14 | |
RDOXTESZEPMUJZ-UHFFFAOYSA-N | |
7519 | |
anisole |
C7H8O | |
MFCD00008354 | |
methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene | |
CHEBI:16579 | |
COC1=CC=CC=C1 |
Specifications
100-66-3 | |
0.994 | |
43°C (109°F) | |
C7H8O | |
MFCD00008354 | |
UN2222 | |
14,669 | |
Miscible with methanol and acetone. Immiscible with water. | |
COC1=CC=CC=C1 | |
108.14 | |
CHEBI:16579 | |
99% |
-37°C | |
153°C to 154°C | |
Ether-like | |
1.517 | |
500 g | |
506892 | |
methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene | |
RDOXTESZEPMUJZ-UHFFFAOYSA-N | |
anisole | |
7519 | |
108.14 | |
Anisole |
Safety and Handling
GHS H Statement
H226-H332-H315-H319-H335
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ANISOLE
EINECSNumber : 202-876-1
RTECSNumber : BZ8050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only